Product Details

SNP ID
rs201779964
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:26239825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATCCAACTGAGGGGATCTCTCA[C/T]GTCACTTCACCGAGTGAGTAGTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP85 PubMed Links

Gene Details

Gene
CEP85
Gene Name
centrosomal protein 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281517.2 222 Silent Mutation CAC,CAT H14H NP_001268446.1
NM_001281518.2 222 Intron NP_001268447.1
NM_001319944.1 222 Silent Mutation CAC,CAT H14H NP_001306873.1
NM_022778.4 222 Silent Mutation CAC,CAT H14H NP_073615.2
XM_017002105.1 222 Silent Mutation CAC,CAT H14H XP_016857594.1
Gene
LOC101928303
Gene Name
uncharacterized LOC101928303
There are no transcripts associated with this gene.

View Full Product Details