Product Details
- SNP ID
-
rs201369951
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:201135778 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGACTGTGTTTGCTCGGGGTCCC[C/T]CGAGGGATGCAGCAGCTGCTGCCAT
- Phenotype
-
MIM: 616877
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM9
PubMed Links
Gene Details
- Gene
- TMEM9
- Gene Name
- transmembrane protein 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001288564.1 |
566 |
Missense Mutation |
GAG,GGG |
E146G |
NP_001275493.1 |
NM_001288565.1 |
566 |
Missense Mutation |
GAG,GGG |
E146G |
NP_001275494.1 |
NM_001288566.1 |
566 |
Missense Mutation |
GAG,GGG |
E146G |
NP_001275495.1 |
NM_001288567.1 |
566 |
Missense Mutation |
GAG,GGG |
E146G |
NP_001275496.1 |
NM_001288568.1 |
566 |
Missense Mutation |
GAG,GGG |
E146G |
NP_001275497.1 |
NM_001288569.1 |
566 |
Missense Mutation |
GAG,GGG |
E146G |
NP_001275498.1 |
NM_001288570.1 |
566 |
Missense Mutation |
GAG,GGG |
E149G |
NP_001275499.1 |
NM_001288571.1 |
566 |
Missense Mutation |
GAG,GGG |
E171G |
NP_001275500.1 |
NM_016456.4 |
566 |
Missense Mutation |
GAG,GGG |
E146G |
NP_057540.1 |
XM_011509385.1 |
566 |
Missense Mutation |
GAG,GGG |
E149G |
XP_011507687.1 |
XM_011509386.2 |
566 |
Missense Mutation |
GAG,GGG |
E149G |
XP_011507688.1 |
XM_011509387.2 |
566 |
Missense Mutation |
GAG,GGG |
E149G |
XP_011507689.1 |
View Full Product Details