Product Details

SNP ID
rs201514015
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:158465952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATATGGCCATCTGTAACCCACTG[C/T]GCTACTCAGTGCTCATGGGACATGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10K1 PubMed Links

Gene Details

Gene
OR10K1
Gene Name
olfactory receptor family 10 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004473.1 391 Missense Mutation CGC,TGC R131C NP_001004473.1

View Full Product Details