Product Details

SNP ID
rs201620424
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:20553324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGAAGCTGACGCTGGGGCCGCA[C/T]GGCATCCGCATGCAGCCGTGCGAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM43B PubMed Links

Gene Details

Gene
FAM43B
Gene Name
family with sequence similarity 43 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207334.2 886 Silent Mutation CAC,CAT H117H NP_997217.1

View Full Product Details