Product Details

SNP ID
rs201739718
Assay Type
Functionally Tested
NCBI dbSNP Submissions
9
Location
Chr.1:203179790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGACGCTTTCCTGGTCGTCGT[A/G]TCCTACCCACTGGTTGCCCTTGGTG
Phenotype
MIM: 601525 MIM: 160795
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CHI3L1 PubMed Links

Gene Details

Gene
CHI3L1
Gene Name
chitinase 3 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276.2 1153 Missense Mutation CAC,TAC H328Y NP_001267.2
Gene
MYBPH
Gene Name
myosin binding protein H
There are no transcripts associated with this gene.

View Full Product Details