Product Details

SNP ID
rs201783745
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:192159285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCTGAGTTTCATGAAGACACCC[G/T]CTCCAGTAGATCTGGGCACTTGGCC
Phenotype
MIM: 607192
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RGS18 PubMed Links

Gene Details

Gene
RGS18
Gene Name
regulator of G-protein signaling 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130782.2 361 Missense Mutation CGC,CTC R62L NP_570138.1
XM_011509891.2 361 Intron XP_011508193.1

View Full Product Details