Product Details
- SNP ID
-
rs201463227
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:151286568 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTGTCAAGAACACTGTGTGTCCC[A/G]AGCAGTCTGAGGCCCTGGCTGGAGG
- Phenotype
-
MIM: 602758
MIM: 610568
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100507670
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2298265] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100507670
- Gene Name
- uncharacterized LOC100507670
There are no transcripts associated with this gene.
- Gene
- PI4KB
- Gene Name
- phosphatidylinositol 4-kinase beta
There are no transcripts associated with this gene.
- Gene
- ZNF687
- Gene Name
- zinc finger protein 687
View Full Product Details