Product Details

SNP ID
rs201541724
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:173907496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATCTGAGACATAGAGGTCATCT[C/T]GGCCTTCTGCAACAATACCTGGAAG
Phenotype
MIM: 107300
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINC1 PubMed Links

Gene Details

Gene
SERPINC1
Gene Name
serpin family C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000488.3 1291 Missense Mutation CAA,CGA Q391R NP_000479.1
XM_005245198.3 1291 Missense Mutation CAA,CGA Q343R XP_005245255.1

View Full Product Details