Product Details

SNP ID
rs201786227
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:9245146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGAGCTGCTCTGACAGCCCCCA[A/G]GCAGGGTCAAGAGCTCATGGCCAAG
Phenotype
MIM: 138090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
H6PD PubMed Links

Gene Details

Gene
H6PD
Gene Name
hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282587.1 485 Missense Mutation AAG,AGG K82R NP_001269516.1
NM_004285.3 485 Missense Mutation AAG,AGG K71R NP_004276.2
XM_005263540.4 485 Missense Mutation AAG,AGG K80R XP_005263597.1
XM_006711052.3 485 Missense Mutation AAG,AGG K71R XP_006711115.1
XM_017002865.1 485 Missense Mutation AAG,AGG K71R XP_016858354.1
XM_017002866.1 485 Intron XP_016858355.1

View Full Product Details