Product Details

SNP ID
rs200974793
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:111230840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAATATTGACCCCTTCCTATGC[C/T]CTCATCTCATCTATTCATTCGCCAG
Phenotype
MIM: 601526
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHI3L2 PubMed Links

Gene Details

Gene
CHI3L2
Gene Name
chitinase 3 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025197.1 240 Missense Mutation CCT,TCT P47S NP_001020368.1
NM_001025199.1 240 UTR 5 NP_001020370.1
NM_004000.2 240 Missense Mutation CCT,TCT P57S NP_003991.2

View Full Product Details