Product Details

SNP ID
rs200987351
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:29148447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGACCTAGATCTAGATCGGGAG[C/G]GCGATCTGGAAGCAGACCTGGATCT
Phenotype
MIM: 601940
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SRSF4 PubMed Links

Gene Details

Gene
SRSF4
Gene Name
serine and arginine rich splicing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005626.4 1531 Missense Mutation CCC,CGC P483R NP_005617.2
XM_011541951.2 1531 Missense Mutation CCC,CGC P453R XP_011540253.1

View Full Product Details