Product Details

SNP ID
rs201186970
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:167055078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGGGCTGCAACCACGCCCACC[A/G]CGATGCCCACATACAGGGCCACGTT
Phenotype
MIM: 602171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPA33 PubMed Links

Gene Details

Gene
GPA33
Gene Name
glycoprotein A33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005814.2 897 Missense Mutation GCG,GTG A242V NP_005805.1
XM_017000005.1 897 Missense Mutation GCG,GTG A134V XP_016855494.1

View Full Product Details