Product Details

SNP ID
rs201515699
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:20114148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGTCTGCCCCCGGCAGTGGGGC[C/T]GCCAGTAGAAACGCAGTCGCTTCTG
Phenotype
MIM: 605630
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G2D PubMed Links

Gene Details

Gene
PLA2G2D
Gene Name
phospholipase A2 group IID
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271814.1 472 UTR 3 NP_001258743.1
NM_012400.3 472 Missense Mutation CAG,CGG Q135R NP_036532.1

View Full Product Details