Product Details

SNP ID
rs201478011
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:149929196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTCAGTGGGATTGAGAATGGTA[C/T]GTGGATCTCTTTTCTTGGAGTGATC
Phenotype
MIM: 605593
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTMR11 PubMed Links

Gene Details

Gene
MTMR11
Gene Name
myotubularin related protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145862.1 3157 Missense Mutation CAT,CGT H688R NP_001139334.1
NM_181873.3 3157 Missense Mutation CAT,CGT H616R NP_870988.2
XM_006711137.1 3157 Missense Mutation CAT,CGT H616R XP_006711200.1
XM_011509099.2 3157 UTR 3 XP_011507401.1
Gene
SF3B4
Gene Name
splicing factor 3b subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005850.4 3157 Intron NP_005841.1

View Full Product Details