Product Details
- SNP ID
-
rs201399564
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:42735965 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCGGGCATGTGCAGCAGAGGAAG[C/G]AGCCGCCCAGCACGGCCAGGCCAGC
- Phenotype
-
MIM: 610036
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN19
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4660658] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLDN19
- Gene Name
- claudin 19
View Full Product Details