Product Details

SNP ID
rs201452471
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:26161404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACGCACCAGGTGATAGCTAGGC[A/G]ACAGGAGCCAGCCCTGCGTGGGAGT
Phenotype
MIM: 616698
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM110D PubMed Links

Gene Details

Gene
FAM110D
Gene Name
family with sequence similarity 110 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024869.2 300 Missense Mutation CAA,CGA Q38R NP_079145.2
Gene
ZNF593
Gene Name
zinc finger protein 593
There are no transcripts associated with this gene.

View Full Product Details