Product Details

SNP ID
rs201242393
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:161191307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGCCAGCCACTAGGACTTGCAGT[G/T]TCAAAGGCTGGGCCAGTGGCCCATG
Phenotype
MIM: 603876 MIM: 602985
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS4 PubMed Links

Gene Details

Gene
ADAMTS4
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320336.1 2610 Missense Mutation GAA,GAC E728D NP_001307265.1
NM_005099.5 2610 Missense Mutation AAA,ACA K782T NP_005090.3
Gene
NDUFS2
Gene Name
NADH:ubiquinone oxidoreductase core subunit S2
There are no transcripts associated with this gene.

View Full Product Details