Product Details

SNP ID
rs201386716
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:12644607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCATCTTTTTCCTGGGGGTCTTT[G/T]TCTGGGCTGTCTTTGAGCACTTCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AADACL4 PubMed Links

Gene Details

Gene
AADACL4
Gene Name
arylacetamide deacetylase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013630.1 61 Missense Mutation GTC,TTC V21F NP_001013652.1
XM_017001152.1 61 Intron XP_016856641.1
XM_017001153.1 61 Intron XP_016856642.1

View Full Product Details