Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001291593.1 | 3288 | Missense Mutation | NP_001278522.1 | ||
NM_001291594.1 | 3288 | Missense Mutation | NP_001278523.1 | ||
NM_015102.4 | 3288 | Missense Mutation | NP_055917.1 | ||
XM_006710563.3 | 3288 | Missense Mutation | XP_006710626.1 | ||
XM_011541213.1 | 3288 | Missense Mutation | XP_011539515.1 | ||
XM_011541214.1 | 3288 | Missense Mutation | XP_011539516.1 | ||
XM_011541215.1 | 3288 | Missense Mutation | XP_011539517.1 | ||
XM_011541216.2 | 3288 | Missense Mutation | XP_011539518.1 | ||
XM_011541217.2 | 3288 | Missense Mutation | XP_011539519.1 | ||
XM_011541218.2 | 3288 | Missense Mutation | XP_011539520.1 | ||
XM_017000996.1 | 3288 | Missense Mutation | XP_016856485.1 | ||
XM_017000997.1 | 3288 | Missense Mutation | XP_016856486.1 | ||
XM_017000998.1 | 3288 | Intron | XP_016856487.1 | ||
XM_017000999.1 | 3288 | Missense Mutation | XP_016856488.1 | ||
XM_017001000.1 | 3288 | Missense Mutation | XP_016856489.1 | ||
XM_017001001.1 | 3288 | Missense Mutation | XP_016856490.1 | ||
XM_017001002.1 | 3288 | Intron | XP_016856491.1 | ||
XM_017001003.1 | 3288 | Missense Mutation | XP_016856492.1 |