Product Details
- SNP ID
-
rs199559320
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:166860274 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCCAAAGGCATATTTAAAATGAC[C/G]ATCTCGTAACCGATAAGCTTTCCTT
- Phenotype
-
MIM: 612763
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
POGK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2272792] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POGK
- Gene Name
- pogo transposable element with KRAB domain
There are no transcripts associated with this gene.
- Gene
- TADA1
- Gene Name
- transcriptional adaptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_053053.3 |
|
Intron |
|
|
NP_444281.1 |
View Full Product Details