Product Details

SNP ID
rs199637318
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:94533131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTCCAATAATAAAGTGTATAAA[C/T]TAAGTCCTTGCCAAAAACATCCCGG
Phenotype
MIM: 134390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F3 PubMed Links

Gene Details

Gene
F3
Gene Name
coagulation factor III, tissue factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178096.1 771 Missense Mutation ATT,GTT I184V NP_001171567.1
NM_001993.4 771 Missense Mutation ATT,GTT I184V NP_001984.1

View Full Product Details