Product Details

SNP ID
rs199651205
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:192575796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCATATTTGCTAAGAGCACCATG[C/G]GCGCAGCAGCCATCTCCACTCCAAA
Phenotype
MIM: 600323
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RGS1 PubMed Links

Gene Details

Gene
RGS1
Gene Name
regulator of G-protein signaling 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002922.3 70 Missense Mutation CGC,GGC R2G NP_002913.3

View Full Product Details