Product Details

SNP ID
rs200114344
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:35560370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGACATAGATGCCAAAACTCG[G/T]CGGCGGATCTTCGATGCTGTCGGCT
Phenotype
MIM: 613535 MIM: 608458
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0319L PubMed Links

Gene Details

Gene
KIAA0319L
Gene Name
KIAA0319 like
There are no transcripts associated with this gene.

Gene
NCDN
Gene Name
neurochondrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014839.1 278 Silent Mutation CGG,CGT R73R NP_001014839.1
NM_001014841.1 278 Silent Mutation CGG,CGT R56R NP_001014841.1
NM_014284.2 278 Silent Mutation CGG,CGT R73R NP_055099.1

View Full Product Details