Product Details

SNP ID
rs199772693
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:23692704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGAGAGTGGAGACAGACTGAC[A/G]CGAGCAGCCAAGGTGTTGGAGCAGC
Phenotype
MIM: 604175
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL11 PubMed Links

Gene Details

Gene
RPL11
Gene Name
ribosomal protein L11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000975.3 147 Silent Mutation NP_000966.2
NM_001199802.1 147 Intron NP_001186731.1

View Full Product Details