Product Details

SNP ID
rs199880094
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:158480198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGATGCTCATCAATCTACTTTC[C/T]GTGGCCAGGACAATCTCCTTCAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10R2 PubMed Links

Gene Details

Gene
OR10R2
Gene Name
olfactory receptor family 10 subfamily R member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004472.1 321 Silent Mutation TCC,TCT S107S NP_001004472.1

View Full Product Details