Product Details
- SNP ID
-
rs199720664
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:91962765 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAAGCAGTTAAGAATGTCTCTGC[C/G]AAGTCGACAAACAGCTATTATTGTT
- Phenotype
-
MIM: 602144
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BRDT
PubMed Links
Gene Details
- Gene
- BRDT
- Gene Name
- bromodomain testis associated
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242805.2 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
NP_001229734.2 |
NM_001242806.2 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
NP_001229735.2 |
NM_001242807.2 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
NP_001229736.2 |
NM_001242808.2 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
NP_001229737.2 |
NM_001242810.2 |
243 |
Intron |
|
|
NP_001229739.2 |
NM_001726.4 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
NP_001717.3 |
NM_207189.3 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
NP_997072.2 |
XM_006710853.3 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
XP_006710916.1 |
XM_006710854.3 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
XP_006710917.1 |
XM_006710855.3 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
XP_006710918.1 |
XM_006710856.3 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
XP_006710919.1 |
XM_006710857.3 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
XP_006710920.1 |
XM_011542032.2 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
XP_011540334.1 |
XM_011542033.2 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
XP_011540335.1 |
XM_011542034.2 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
XP_011540336.1 |
XM_011542035.2 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
XP_011540337.1 |
XM_011542036.2 |
243 |
Missense Mutation |
CCA,CGA |
P4R |
XP_011540338.1 |
View Full Product Details