Product Details

SNP ID
rs199772818
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:28564772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATGGCATGCTGTATGAATTCTT[C/T]GTCAAAGTCTACCCCTCCTGTCGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRNAU1AP PubMed Links

Gene Details

Gene
TRNAU1AP
Gene Name
tRNA selenocysteine 1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017846.4 616 Silent Mutation TTC,TTT F116F NP_060316.1
XM_017001542.1 616 Silent Mutation TTC,TTT F6F XP_016857031.1

View Full Product Details