Product Details
- SNP ID
-
rs200026035
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:151133140 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTCCCAGGGGTCCCCGGCGGCGC[A/G]GAGGTGCTTGACCGGCAGCTCCGGC
- Phenotype
-
MIM: 609294
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GABPB2
PubMed Links
Gene Details
- Gene
- GABPB2
- Gene Name
- GA binding protein transcription factor beta subunit 2
There are no transcripts associated with this gene.
- Gene
- SEMA6C
- Gene Name
- semaphorin 6C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001178061.1 |
3449 |
Missense Mutation |
CGC,TGC |
R745C |
NP_001171532.1 |
NM_001178062.1 |
3449 |
Missense Mutation |
CGC,TGC |
R705C |
NP_001171533.1 |
NM_030913.4 |
3449 |
Missense Mutation |
CGC,TGC |
R713C |
NP_112175.2 |
XM_005244835.3 |
3449 |
Missense Mutation |
CGC,TGC |
R745C |
XP_005244892.1 |
XM_017000075.1 |
3449 |
Missense Mutation |
CGC,TGC |
R745C |
XP_016855564.1 |
XM_017000076.1 |
3449 |
Missense Mutation |
CGC,TGC |
R745C |
XP_016855565.1 |
XM_017000077.1 |
3449 |
Missense Mutation |
CGC,TGC |
R745C |
XP_016855566.1 |
XM_017000078.1 |
3449 |
Missense Mutation |
CGC,TGC |
R713C |
XP_016855567.1 |
XM_017000079.1 |
3449 |
Missense Mutation |
CGC,TGC |
R713C |
XP_016855568.1 |
XM_017000080.1 |
3449 |
Missense Mutation |
CGC,TGC |
R705C |
XP_016855569.1 |
XM_017000081.1 |
3449 |
Missense Mutation |
CGC,TGC |
R705C |
XP_016855570.1 |
XM_017000082.1 |
3449 |
Missense Mutation |
CGC,TGC |
R480C |
XP_016855571.1 |
View Full Product Details