Product Details

SNP ID
rs193920807
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:158699706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATATCTTACTTGAGGTCCCTGGT[C/A]TTACGGAAAAAAATATCTTAGCTTG
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
OR6K2 PubMed Links

Gene Details

Gene
OR6K2
Gene Name
olfactory receptor family 6 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005279.1 947 Missense Mutation AGA,ATA R316I NP_001005279.1

View Full Product Details