Product Details

SNP ID
rs199681084
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:231224289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGACAAAGTACTTCATTTGGCC[A/G]CTCTGGAAGAATTTATTTTCTTGAT
Phenotype
MIM: 610584
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf131 PubMed Links

Gene Details

Gene
C1orf131
Gene Name
chromosome 1 open reading frame 131
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300830.1 918 Missense Mutation GCG,GTG A290V NP_001287759.1
NM_152379.3 918 Missense Mutation GCG,GTG A291V NP_689592.2
Gene
TRIM67
Gene Name
tripartite motif containing 67
There are no transcripts associated with this gene.

View Full Product Details