Product Details

SNP ID
rs199509938
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:23794274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCACCTGCCCCCACCCTCTGGC[C/T]GGCATCGTGGCGTTGAGCTGCTGGC
Phenotype
MIM: 606953 MIM: 613898 MIM: 616143
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALE PubMed Links

Gene Details

Gene
GALE
Gene Name
UDP-galactose-4-epimerase
There are no transcripts associated with this gene.

Gene
HMGCL
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase
There are no transcripts associated with this gene.

Gene
LYPLA2
Gene Name
lysophospholipase II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007260.2 908 Intron NP_009191.1
XM_005245728.4 908 Silent Mutation GCC,GCT A152A XP_005245785.1
XM_017000165.1 908 Silent Mutation GCC,GCT A140A XP_016855654.1
Gene
PITHD1
Gene Name
PITH domain containing 1
There are no transcripts associated with this gene.

View Full Product Details