Product Details

SNP ID
rs199575981
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:220102412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGTTTTGGTCTCCGTCATCTA[C/G]GTATATATGCATTTTTCGGTAATTA
Phenotype
MIM: 612801
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IARS2 PubMed Links

Gene Details

Gene
IARS2
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018060.3 853 Missense Mutation ACG,AGG T250R NP_060530.3

View Full Product Details