Product Details

SNP ID
rs199655500
Assay Type
Functionally Tested
NCBI dbSNP Submissions
16
Location
Chr.1:201900333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTTCTGGTGTCTGTGTTCCTA[C/T]GCTCCCCTTTTACCTTCTCATCATC
Phenotype
MIM: 602715
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LMOD1 PubMed Links

Gene Details

Gene
LMOD1
Gene Name
leiomodin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012134.2 928 Missense Mutation NP_036266.2
Gene
SHISA4
Gene Name
shisa family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198149.2 928 Intron NP_937792.2
XM_017000419.1 928 Intron XP_016855908.1

View Full Product Details