Product Details

SNP ID
rs200002263
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:247712056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCACGTGGAGAAGGCTTTGCTCC[A/G]GCCACTGGCAGAGGGGATCCTGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6F1 PubMed Links

Gene Details

Gene
OR6F1
Gene Name
olfactory receptor family 6 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005286.1 700 Missense Mutation CGG,TGG R234W NP_001005286.1

View Full Product Details