Product Details

SNP ID
rs199577888
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:153995093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCTACCTAGGAAGATGGAAGCT[A/G]TTGATGCCAGCACTGCAAAGAGGGT
Phenotype
MIM: 602672 MIM: 603702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUP210L PubMed Links

Gene Details

Gene
NUP210L
Gene Name
nucleoporin 210 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159484.1 5546 Missense Mutation ACA,ATA T1673I NP_001152956.1
NM_207308.2 5546 Missense Mutation ACA,ATA T1825I NP_997191.2
XM_011510122.1 5546 Missense Mutation ACA,ATA T1781I XP_011508424.1
XM_011510123.1 5546 Missense Mutation ACA,ATA T1772I XP_011508425.1
XM_011510124.1 5546 UTR 3 XP_011508426.1
XM_017002788.1 5546 Intron XP_016858277.1
XM_017002789.1 5546 Intron XP_016858278.1
Gene
RAB13
Gene Name
RAB13, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
RPS27
Gene Name
ribosomal protein S27
There are no transcripts associated with this gene.

View Full Product Details