Product Details
- SNP ID
-
rs199737051
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:161509829 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTTGTGTCTTTCAGAATGGCTGG[G/T]GCTCCAGACCCCTCACCTGGAGTTC
- Phenotype
-
MIM: 146790
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FCGR2A
PubMed Links
Gene Details
- Gene
- FCGR2A
- Gene Name
- Fc fragment of IgG receptor IIa
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001136219.1 |
412 |
Missense Mutation |
GGG,GTG |
G125V |
NP_001129691.1 |
NM_021642.3 |
412 |
Missense Mutation |
GGG,GTG |
G124V |
NP_067674.2 |
XM_011509287.2 |
412 |
Missense Mutation |
GGG,GTG |
G125V |
XP_011507589.1 |
XM_011509290.2 |
412 |
Missense Mutation |
GGG,GTG |
G125V |
XP_011507592.1 |
XM_011509291.1 |
412 |
Missense Mutation |
GGG,GTG |
G125V |
XP_011507593.1 |
XM_017000663.1 |
412 |
Missense Mutation |
GGG,GTG |
G124V |
XP_016856152.1 |
XM_017000664.1 |
412 |
Missense Mutation |
GGG,GTG |
G125V |
XP_016856153.1 |
XM_017000665.1 |
412 |
Missense Mutation |
GGG,GTG |
G125V |
XP_016856154.1 |
XM_017000666.1 |
412 |
Missense Mutation |
GGG,GTG |
G125V |
XP_016856155.1 |
XM_017000667.1 |
412 |
Missense Mutation |
GGG,GTG |
G125V |
XP_016856156.1 |
XM_017000668.1 |
412 |
Missense Mutation |
GGG,GTG |
G124V |
XP_016856157.1 |
View Full Product Details