Product Details

SNP ID
rs200003317
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:83870002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGATGTAAACAAAACATTGGAG[C/T]GAGTCATTCCAGGTGAATTGTACTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927560 PubMed Links

Gene Details

Gene
LOC101927560
Gene Name
uncharacterized LOC101927560
There are no transcripts associated with this gene.

Gene
LOC101927587
Gene Name
uncharacterized LOC101927587
There are no transcripts associated with this gene.

Gene
TTLL7
Gene Name
tubulin tyrosine ligase like 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024686.4 2795 Missense Mutation CAC,CGC H875R NP_078962.4
XM_005271208.2 2795 Missense Mutation CAC,CGC H848R XP_005271265.1
XM_011542159.2 2795 Intron XP_011540461.1
XM_017002347.1 2795 Missense Mutation CAC,CGC H875R XP_016857836.1
XM_017002348.1 2795 Intron XP_016857837.1
XM_017002349.1 2795 Missense Mutation CAC,CGC H630R XP_016857838.1

View Full Product Details