Product Details

SNP ID
rs199671097
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:247896030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCCGGGGCTTGGTGTCAGTGAC[C/G]TGGGGCTGTGGGGTGGCCAACTCCT
Phenotype
MIM: 616729
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR2W3 PubMed Links

Gene Details

Gene
OR2W3
Gene Name
olfactory receptor family 2 subfamily W member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001957.2 444 Silent Mutation ACC,ACG T148T NP_001001957.2

View Full Product Details