Product Details

SNP ID
rs199684527
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:155256742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGGCGACCAGCAGCATGAGCA[C/T]GGATACTGCTGTGTTGCCCTTCGGC
Phenotype
MIM: 602989 MIM: 606913
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLK2 PubMed Links

Gene Details

Gene
CLK2
Gene Name
CDC like kinase 2
There are no transcripts associated with this gene.

Gene
FAM189B
Gene Name
family with sequence similarity 189 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267608.1 985 Intron NP_001254537.1
NM_006589.2 985 Intron NP_006580.2
NM_198264.1 985 Intron NP_937995.1
Gene
SCAMP3
Gene Name
secretory carrier membrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005698.3 985 Missense Mutation ATG,GTG M277V NP_005689.2
NM_052837.2 985 Missense Mutation ATG,GTG M251V NP_443069.1
XM_006711105.3 985 Missense Mutation ATG,GTG M263V XP_006711168.1
XM_006711106.3 985 Missense Mutation ATG,GTG M210V XP_006711169.1
XM_016999991.1 985 Silent Mutation CCA,CCG P189P XP_016855480.1

View Full Product Details