Product Details

SNP ID
rs199737525
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:151851084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCTCCAGGTAGGAGCCTGGTT[A/G]GAAAAGACAGACCGACTTCTTCTGG
Phenotype
MIM: 615653
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100132111 PubMed Links

Gene Details

Gene
LOC100132111
Gene Name
uncharacterized LOC100132111
There are no transcripts associated with this gene.

Gene
THEM5
Gene Name
thioesterase superfamily member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182578.3 565 Nonsense Mutation CAA,TAA Q145* NP_872384.1
XM_011509421.1 565 Nonsense Mutation CAA,TAA Q145* XP_011507723.1

View Full Product Details