Product Details

SNP ID
rs200088823
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:28742621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCCTAGGTAGCACTCCATTTCT[C/T]GGTCAGCATGGTTTTAATTTCTTTC
Phenotype
MIM: 610640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YTHDF2 PubMed Links

Gene Details

Gene
YTHDF2
Gene Name
YTH N6-methyladenosine RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172828.1 848 Silent Mutation CTC,CTT L67L NP_001166299.1
NM_001173128.1 848 Silent Mutation CTC,CTT L117L NP_001166599.1
NM_016258.2 848 Silent Mutation CTC,CTT L117L NP_057342.2

View Full Product Details