Product Details

SNP ID
rs199500317
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:153149891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCTGATGAATTCTAGTGATGTT[A/G]CTTGCTCTTGGGTGGATACTTCTGC
Phenotype
MIM: 182269
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPRR2C PubMed Links

Gene Details

Gene
SPRR2C
Gene Name
small proline rich protein 2C (pseudogene)
There are no transcripts associated with this gene.

Gene
SPRR2G
Gene Name
small proline rich protein 2G
There are no transcripts associated with this gene.

View Full Product Details