Product Details

SNP ID
rs200090728
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:247815451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAAAGACCTGGGAAACACAGCC[A/T]AGGAATGAAATGGAGTTGTTGTGTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR14A16 PubMed Links

Gene Details

Gene
OR14A16
Gene Name
olfactory receptor family 14 subfamily A member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001966.1 279 Silent Mutation CTA,CTT L93L NP_001001966.1

View Full Product Details