Product Details

SNP ID
rs200092801
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:46673367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTCCTATCCTTTGGCCACCTGT[C/T]CTGAAAGGAATGTTCTCTTCCATTC
Phenotype
MIM: 608917
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATPAF1 PubMed Links

Gene Details

Gene
ATPAF1
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
There are no transcripts associated with this gene.

Gene
EFCAB14
Gene Name
EF-hand calcium binding domain 14
There are no transcripts associated with this gene.

Gene
EFCAB14-AS1
Gene Name
EFCAB14 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TEX38
Gene Name
testis expressed 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145474.3 634 Missense Mutation CCT,TCT P178S NP_001138946.1
NM_001300863.1 634 Missense Mutation CCT,TCT P124S NP_001287792.1
NM_001300864.1 634 Missense Mutation CCT,TCT P102S NP_001287793.1
XM_011541421.2 634 Missense Mutation CCT,TCT P179S XP_011539723.1

View Full Product Details