Product Details

SNP ID
rs199554966
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:159855995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAGCCCCCGCAGCAGCAGCAGA[C/G]GAGCCCCCAGATGCCTACGGCCAGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf204 PubMed Links

Gene Details

Gene
C1orf204
Gene Name
chromosome 1 open reading frame 204
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134233.1 995 Intron NP_001127705.1
Gene
VSIG8
Gene Name
V-set and immunoglobulin domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013661.1 995 Missense Mutation CTC,GTC L287V NP_001013683.1

View Full Product Details