Product Details
- SNP ID
-
rs199766172
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:245749628 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATCAAATCTTCAATCAGGCTGT[A/G]TTCTCTGCCATGTGTCACTCTCATA
- Phenotype
-
MIM: 608783
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SMYD3
PubMed Links
Gene Details
- Gene
- SMYD3
- Gene Name
- SET and MYND domain containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001167740.1 |
1175 |
Missense Mutation |
CAC,TAC |
H408Y |
NP_001161212.1 |
NM_022743.2 |
1175 |
Missense Mutation |
CAC,TAC |
H349Y |
NP_073580.1 |
XM_011544253.1 |
1175 |
Missense Mutation |
CAC,TAC |
H371Y |
XP_011542555.1 |
XM_011544254.2 |
1175 |
Missense Mutation |
CAC,TAC |
H266Y |
XP_011542556.1 |
XM_011544257.1 |
1175 |
Missense Mutation |
CAC,TAC |
H238Y |
XP_011542559.1 |
XM_011544258.2 |
1175 |
Missense Mutation |
CAC,TAC |
H219Y |
XP_011542560.1 |
XM_011544259.2 |
1175 |
Missense Mutation |
CAC,TAC |
H219Y |
XP_011542561.1 |
XM_011544260.1 |
1175 |
Missense Mutation |
CAC,TAC |
H219Y |
XP_011542562.1 |
XM_011544261.2 |
1175 |
Missense Mutation |
CAC,TAC |
H219Y |
XP_011542563.1 |
XM_017002094.1 |
1175 |
Missense Mutation |
CAC,TAC |
H219Y |
XP_016857583.1 |
XM_017002095.1 |
1175 |
Missense Mutation |
CAC,TAC |
H216Y |
XP_016857584.1 |
View Full Product Details