Product Details

SNP ID
rs199927668
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:6616847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCCTATGCTGGCTACATCCCT[C/T]ATCCGAAGGAGGTAATCTTCTGAGT
Phenotype
MIM: 612532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHF13 PubMed Links

Gene Details

Gene
PHF13
Gene Name
PHD finger protein 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153812.2 501 Missense Mutation CAT,TAT H44Y NP_722519.2
XM_011540762.1 501 Missense Mutation CAT,TAT H53Y XP_011539064.1
Gene
THAP3
Gene Name
THAP domain containing 3
There are no transcripts associated with this gene.

View Full Product Details