Product Details

SNP ID
rs200096396
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:93154797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGACTCTATCAAAGTTGCTTTCT[C/T]GTATGTTTCGATCACGAGCTTCAAA
Phenotype
MIM: 616876
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMED5 PubMed Links

Gene Details

Gene
TMED5
Gene Name
transmembrane p24 trafficking protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167830.1 1059 UTR 3 NP_001161302.1
NM_016040.4 1059 Missense Mutation CAA,CGA Q188R NP_057124.3

View Full Product Details