Product Details
- SNP ID
-
rs200019262
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:84647580 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACCTGATTTGGTTTAATTTCTTC[A/G]GCAGTTATATTCAGTACATTGATTG
- Phenotype
-
MIM: 608690
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SSX2IP
PubMed Links
Gene Details
- Gene
- SSX2IP
- Gene Name
- SSX family member 2 interacting protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001166293.1 |
1974 |
Silent Mutation |
GCC,GCT |
A566A |
NP_001159765.1 |
NM_001166294.1 |
1974 |
Silent Mutation |
GCC,GCT |
A539A |
NP_001159766.1 |
NM_001166295.1 |
1974 |
Silent Mutation |
GCC,GCT |
A539A |
NP_001159767.1 |
NM_001166417.1 |
1974 |
Silent Mutation |
GCC,GCT |
A566A |
NP_001159889.1 |
NM_014021.3 |
1974 |
Silent Mutation |
GCC,GCT |
A566A |
NP_054740.3 |
XM_005270427.1 |
1974 |
Silent Mutation |
GCC,GCT |
A566A |
XP_005270484.1 |
XM_005270428.1 |
1974 |
Silent Mutation |
GCC,GCT |
A566A |
XP_005270485.1 |
XM_005270429.1 |
1974 |
Silent Mutation |
GCC,GCT |
A565A |
XP_005270486.1 |
XM_006710331.1 |
1974 |
Silent Mutation |
GCC,GCT |
A539A |
XP_006710394.1 |
XM_011540610.1 |
1974 |
Silent Mutation |
GCC,GCT |
A566A |
XP_011538912.1 |
XM_011540612.2 |
1974 |
UTR 3 |
|
|
XP_011538914.1 |
XM_017000234.1 |
1974 |
Silent Mutation |
GCC,GCT |
A566A |
XP_016855723.1 |
XM_017000235.1 |
1974 |
Silent Mutation |
GCC,GCT |
A565A |
XP_016855724.1 |
XM_017000236.1 |
1974 |
Silent Mutation |
GCC,GCT |
A565A |
XP_016855725.1 |
XM_017000237.1 |
1974 |
Silent Mutation |
GCC,GCT |
A538A |
XP_016855726.1 |
View Full Product Details