Product Details
- SNP ID
-
rs199794109
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:11858720 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTCACCTGTAACCCGGACGTTTC[C/T]AAGTCCGAGGCTGAACCGGGGCTGC
- Phenotype
-
MIM: 600295
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NPPB
PubMed Links
Gene Details
- Gene
- NPPB
- Gene Name
- natriuretic peptide B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002521.2 |
216 |
Silent Mutation |
TTA,TTG |
L38L |
NP_002512.1 |
View Full Product Details